Anti-GSTA1 Mouse Monoclonal Antibody [clone: [2F7]]

Supplier: Prosci

51-953
10077-316EA 647.66 USD
10077-316
Anti-GSTA1 Mouse Monoclonal Antibody [clone: [2F7]]
Antibodies
GSTA1 antibody can be used in Western Blot starting at 1:500, and immunohistochemistry starting at 1:250.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GSTA1
Clonality: Monoclonal
Clone: [2F7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR