Anti-FOXO3 Mouse Monoclonal Antibody [clone: [4F2]]

51-949
10077-308EA 836.5 USD
10077-308
Anti-FOXO3 Mouse Monoclonal Antibody [clone: [4F2]]
Antibodies
FOXO3 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXO3
Clonality: Monoclonal
Clone: [4F2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR