Anti-FOXO3 Mouse Monoclonal Antibody [clone: [2H2]]

Supplier: Prosci

51-947
10077-304EA 647.66 USD
10077-304
Anti-FOXO3 Mouse Monoclonal Antibody [clone: [2H2]]
Antibodies
FOXO3 antibody can be used in ELISA starting at 1:100 - 1:1000, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXO3
Clonality: Monoclonal
Clone: [2H2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR