Anti-DLD Mouse Monoclonal Antibody [clone: [1G11]]

Supplier: Prosci

51-939
10079-640EA 647.66 USD
10079-640
Anti-DLD Mouse Monoclonal Antibody [clone: [1G11]]
Antibodies
DLDH antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry, and immunoprecipitation.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLD
Clonality: Monoclonal
Clone: [1G11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR