Anti-CSTB Mouse Monoclonal Antibody [clone: [M1-C1]]

51-935
10077-296EA 760.5 USD
10077-296
Anti-CSTB Mouse Monoclonal Antibody [clone: [M1-C1]]
Antibodies
Stefin B antibody can be used in immunohistochemistry starting at 1:50, and flow cytometry.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CSTB
Clonality: Monoclonal
Clone: [M1-C1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR