Anti-TLR4 Mouse Monoclonal Antibody [clone: [3B6]]

51-929
10077-286EA 749.47 USD
10077-286
Anti-TLR4 Mouse Monoclonal Antibody [clone: [3B6]]
Antibodies
TLR4 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

TLR4 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: [3B6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR