Anti-TG Mouse Monoclonal Antibody

51-919
10073-498EA 540.97 USD
10073-498
Anti-TG Mouse Monoclonal Antibody
Thyroglobulin antibody can be used in Western Blot starting at 5 - 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Thyroglobulin antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR