Anti-CD44 Mouse Monoclonal Antibody [clone: [8E2F3]]

51-918
10073-496EA 684.46 USD
10073-496
Anti-CD44 Mouse Monoclonal Antibody [clone: [8E2F3]]
Antibodies
CD44 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: [8E2F3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR