Anti-ATF5 Mouse Monoclonal Antibody [clone: [4G5]]

51-908
10073-476EA 760.5 USD
10073-476
Anti-ATF5 Mouse Monoclonal Antibody [clone: [4G5]]
Antibodies
ATF5 antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

ATF5 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ATF5
Clonality: Monoclonal
Clone: [4G5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR