Anti-OLIG2 Mouse Monoclonal Antibody [clone: [3C9]]

51-906
10077-280EA 778.14 USD
10077-280
Anti-OLIG2 Mouse Monoclonal Antibody [clone: [3C9]]
Antibodies
OLIG2 antibody can be used in immunohistochemistry starting at 5 ?g/mL, and flow cytometry.

PBS, pH 7.2

OLIG2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: OLIG2
Clonality: Monoclonal
Clone: [3C9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR