Anti-CREB1 Mouse Monoclonal Antibody [clone: [5F2]]

51-903
10073-474EA 749.47 USD
10073-474
Anti-CREB1 Mouse Monoclonal Antibody [clone: [5F2]]
Antibodies
CREB1 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry starting at 1 µg/1E6 cells.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CREB1
Clonality: Monoclonal
Clone: [5F2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR