Anti-CCNH Mouse Monoclonal Antibody [clone: [1B8]]

Supplier: ProSci
51-897
10073-462EA 749.47 USD
10073-462
Anti-CCNH Mouse Monoclonal Antibody [clone: [1B8]]
Antibodies
Cyclin H antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry starting at 1 µg/1E6 cells.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCNH
Clonality: Monoclonal
Clone: [1B8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR