Anti-ASAH1 Mouse Monoclonal Antibody [clone: [1A7]]

Supplier: Prosci

51-886
10079-624EA 647.66 USD
10079-624
Anti-ASAH1 Mouse Monoclonal Antibody [clone: [1A7]]
Antibodies
ASAH1 antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ASAH1
Clonality: Monoclonal
Clone: [1A7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR