Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: [1G6]]

Supplier: Prosci

51-880
10073-452EA 647.66 USD
10073-452
Anti-ALDH1A1 Mouse Monoclonal Antibody [clone: [1G6]]
Antibodies
ALDH1A1 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALDH1A1
Clonality: Monoclonal
Clone: [1G6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR