Anti-ACTA2 Mouse Monoclonal Antibody [clone: [1D11-2B9]]

Supplier: Prosci

51-877
10073-446EA 647.66 USD
10073-446
Anti-ACTA2 Mouse Monoclonal Antibody [clone: [1D11-2B9]]
Antibodies
ACTA2 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store ACTA2 antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: [1D11-2B9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR