Anti-ICAM3 Mouse Monoclonal Antibody [clone: [2F8]]

Supplier: ProSci
51-866
10073-430EA 749.47 USD
10073-430
Anti-ICAM3 Mouse Monoclonal Antibody [clone: [2F8]]
Antibodies
ICAM3 antibody can be used in Western Blot, immunohistochemistry starting at 20 ?g/mL, and immunoprecipitation.

PBS, pH 7.2

Store ICAM3 antibody at -20 °C or lower. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: [2F8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR