Anti-F10 Sheep Polyclonal Antibody

Supplier: Prosci

51-860
10073-418EA 582.9 USD
10073-418
Anti-F10 Sheep Polyclonal Antibody
Antibodies
Factor 10 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

10 mM HEPES, pH 7.4, 0.15 M NaCl, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR