Anti-F13A1 Sheep Polyclonal Antibody

Supplier: Prosci

51-856
10073-412EA 582.9 USD
10073-412
Anti-F13A1 Sheep Polyclonal Antibody
Antibodies
Factor 13A1 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

0.01 M HEPES, pH 7.4, 0.15 M NaCl, 50% glycerol. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: F13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR