Anti-EGFR Sheep Polyclonal Antibody

Supplier: Prosci

51-852
10073-404EA 582.9 USD
10073-404
Anti-EGFR Sheep Polyclonal Antibody
Antibodies
EGFR antibody can be used in Western Blot starting at 1:250 - 1:500, immunohistochemistry starting at 5 ?g/mL, and immunocytochemistry starting at 1:100 - 1:200.

PBS buffer pH 7.4

Store EGFR antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR