Anti-KLKB1 Sheep Polyclonal Antibody

Supplier: ProSci
51-848
10073-396EA 674.53 USD
10073-396
Anti-KLKB1 Sheep Polyclonal Antibody
Antibodies
KLKB1 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

10 mM HEPES, pH 7.4, 0.15 M NaCl, 50% glycerol.

Store KLKB1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: KLKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR