Anti-CD59 Rat Monoclonal Antibody

Supplier: Prosci

51-845
10073-392EA 582.9 USD
10073-392
Anti-CD59 Rat Monoclonal Antibody
Antibodies
CD59 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.09% sodium azide.

Store CD59 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR