Anti-C5AR1 Rat Monoclonal Antibody

Supplier: ProSci
51-844
10073-390EA 698.16 USD
10073-390
Anti-C5AR1 Rat Monoclonal Antibody
Antibodies
C5AR antibody can be used in Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 1:100.

PBS, pH 7.4, 0.09% sodium azide.

Store C5AR antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: C5AR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR