Anti-IL10 Rat Monoclonal Antibody [clone: [JES5-2A5]]

Supplier: ProSci
51-843
10073-388EA 698.16 USD
10073-388
Anti-IL10 Rat Monoclonal Antibody [clone: [JES5-2A5]]
Antibodies
IL-10 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.4, 0.09% sodium azide.

Store IL-10 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: [JES5-2A5]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR