Anti-BID Rat Monoclonal Antibody [clone: [1H11]]

Supplier: Prosci

51-837
10073-376EA 582.9 USD
10073-376
Anti-BID Rat Monoclonal Antibody [clone: [1H11]]
Antibodies
BID antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 ?g/mL.

Aqueous buffer, 0.09% sodium azide, contains stabilizer if necessary

Store BID antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type: Primary
Antigen: BID
Clonality: Monoclonal
Clone: [1H11]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR