Anti-YY1 Mouse Monoclonal Antibody [clone: [4D2]]

Supplier: Prosci

51-820
10079-594EA 647.66 USD
10079-594
Anti-YY1 Mouse Monoclonal Antibody [clone: [4D2]]
Antibodies
YY1 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, immunohistochemistry starting at 2.5 ?g/mL, and immunoprecipitation.

PBS, pH 7.2

YY1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: YY1
Clonality: Monoclonal
Clone: [4D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR