Anti-NR2C2 Mouse Monoclonal Antibody [clone: [2A5]]

Supplier: ProSci
51-815
10079-584EA 749.47 USD
10079-584
Anti-NR2C2 Mouse Monoclonal Antibody [clone: [2A5]]
Antibodies
NR2C2 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

NR2C2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: NR2C2
Clonality: Monoclonal
Clone: [2A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR