Anti-SFPQ Mouse Monoclonal Antibody [clone: [6D7]]

Supplier: ProSci
51-799
10077-272EA 749.47 USD
10077-272
Anti-SFPQ Mouse Monoclonal Antibody [clone: [6D7]]
Antibodies
SFPQ antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

PBS, pH 7.2

SFPQ antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SFPQ
Clonality: Monoclonal
Clone: [6D7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR