Anti-CXCL5 Mouse Monoclonal Antibody [clone: [2A9]]

Supplier: ProSci
51-798
10077-270EA 798.53 USD
10077-270
Anti-CXCL5 Mouse Monoclonal Antibody [clone: [2A9]]
Antibodies
CXCL5 antibody can be used in Western Blot at 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation starting at 2 ?g/mL.

PBS, pH 7.2

CXCL5 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CXCL5
Clonality: Monoclonal
Clone: [2A9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR