Anti-S100 A6 Mouse Monoclonal Antibody [clone: [6B5]]

Supplier: ProSci
51-797
10077-268EA 749.47 USD
10077-268
Anti-S100 A6 Mouse Monoclonal Antibody [clone: [6B5]]
Antibodies
S100A6 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry, and immunoprecipitation.

PBS, pH 7.2

S100A6 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: S100A6
Clonality: Monoclonal
Clone: [6B5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR