Anti-RBBP4 Mouse Monoclonal Antibody

Supplier: Prosci

51-795
10077-264EA 624.31 USD InStock
10077-264
Anti-RBBP4 Mouse Monoclonal Antibody
Antibodies
RBAP48 antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, immunocytochemistry, and immunoprecipitation

PBS, pH 7.2

RBAP48 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: RBBP4
Clonality: Monoclonal
Clone: [2D7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR