Anti-PIM1 Mouse Monoclonal Antibody [clone: [2C9]]

Supplier: ProSci
51-780
10074-890EA 749.47 USD
10074-890
Anti-PIM1 Mouse Monoclonal Antibody [clone: [2C9]]
Antibodies
PIM1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunocytochemistry.

PBS, pH 7.2

PIM1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PIM1
Clonality: Monoclonal
Clone: [2C9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR