Anti-LHX4 Mouse Monoclonal Antibody [clone: [1D6]]

Supplier: ProSci
51-772
10074-874EA 749.47 USD
10074-874
Anti-LHX4 Mouse Monoclonal Antibody [clone: [1D6]]
Antibodies
LHX4 antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, pH 7.2

LHX4 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: LHX4
Clonality: Monoclonal
Clone: [1D6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR