Anti-FBXO11 Mouse Monoclonal Antibody [clone: [4C12]]

Supplier: Prosci

51-768
10074-866EA 647.66 USD
10074-866
Anti-FBXO11 Mouse Monoclonal Antibody [clone: [4C12]]
Antibodies
FBXO11 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

FBXO11 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: FBXO11
Clonality: Monoclonal
Clone: [4C12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR