Anti-JAM2 Mouse Monoclonal Antibody [clone: [1G4]]

Supplier: ProSci
51-766
10074-862EA 749.47 USD
10074-862
Anti-JAM2 Mouse Monoclonal Antibody [clone: [1G4]]
Antibodies
JAMB antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

JAMB antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: JAM2
Clonality: Monoclonal
Clone: [1G4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR