Anti-SMURF1 Mouse Monoclonal Antibody [clone: [1D7]]

Supplier: ProSci
51-764
10074-858EA 836.5 USD
10074-858
Anti-SMURF1 Mouse Monoclonal Antibody [clone: [1D7]]
Antibodies
SMURF1 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

SMURF1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SMURF1
Clonality: Monoclonal
Clone: [1D7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR