Anti-MEF2A Mouse Monoclonal Antibody [clone: [3F6]]

Supplier: Prosci

51-759
10074-848EA 647.66 USD
10074-848
Anti-MEF2A Mouse Monoclonal Antibody [clone: [3F6]]
Antibodies
MEF2A antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

MEF2A antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: MEF2A
Clonality: Monoclonal
Clone: [3F6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR