Anti-CD46 Mouse Monoclonal Antibody [clone: [MEM-258]]

Supplier: Prosci

51-756
10074-842EA 582.9 USD
10074-842
Anti-CD46 Mouse Monoclonal Antibody [clone: [MEM-258]]
Antibodies
CD46 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, 0.09% sodium azide.

CD46 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: [MEM-258]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR