Anti-HSPB8 Mouse Monoclonal Antibody [clone: [5B12]]

Supplier: ProSci
51-751
10077-240EA 778.14 USD
10077-240
Anti-HSPB8 Mouse Monoclonal Antibody [clone: [5B12]]
Antibodies
HSPB8 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

HSPB8 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: [5B12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR