Anti-PHGDH Mouse Monoclonal Antibody [clone: [4A3-1D6]]

Supplier: Prosci

51-750
10077-238EA 647.66 USD
10077-238
Anti-PHGDH Mouse Monoclonal Antibody [clone: [4A3-1D6]]
Antibodies
PHGDH antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

PHGDH antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PHGDH
Clonality: Monoclonal
Clone: [4A3-1D6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR