Anti-MKNK1 Mouse Monoclonal Antibody [clone: [2F12]]

Supplier: Prosci

51-732
10077-202EA 647.66 USD
10077-202
Anti-MKNK1 Mouse Monoclonal Antibody [clone: [2F12]]
Antibodies
MNK1 antibody can be used in Western Blot starting at 2 - 4 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

MNK1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: MKNK1
Clonality: Monoclonal
Clone: [2F12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR