Anti-GALT Mouse Monoclonal Antibody [clone: [4C11]]

Supplier: ProSci
51-706
10073-334EA 778.14 USD
10073-334
Anti-GALT Mouse Monoclonal Antibody [clone: [4C11]]
Antibodies
GALT antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

GALT antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: GALT
Clonality: Monoclonal
Clone: [4C11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR