Anti-ENO3 Mouse Monoclonal Antibody [clone: [5D1]]

Supplier: ProSci
51-698
10073-320EA 749.47 USD
10073-320
Anti-ENO3 Mouse Monoclonal Antibody [clone: [5D1]]
Antibodies
ENO3 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

ENO3 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ENO3
Clonality: Monoclonal
Clone: [5D1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR