Anti-EN1 Mouse Monoclonal Antibody [clone: [3H2]]

Supplier: ProSci
51-696
10073-316EA 749.47 USD
10073-316
Anti-EN1 Mouse Monoclonal Antibody [clone: [3H2]]
Antibodies
EN1 antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

EN1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: EN1
Clonality: Monoclonal
Clone: [3H2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR