Anti-DLX2 Mouse Monoclonal Antibody [clone: [2E12]]

Supplier: ProSci
51-693
10073-310EA 749.47 USD
10073-310
Anti-DLX2 Mouse Monoclonal Antibody [clone: [2E12]]
Antibodies
DLX2 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

DLX2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: DLX2
Clonality: Monoclonal
Clone: [2E12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR