Anti-TPP1 Mouse Monoclonal Antibody

Supplier: ProSci
51-687
10074-834EA 589.48 USD
10074-834
Anti-TPP1 Mouse Monoclonal Antibody
TPP1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

TPP1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: TPP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR