Anti-SERPINA3 Mouse Monoclonal Antibody [clone: [1E6]]

Supplier: ProSci
51-667
10079-578EA 786.95 USD
10079-578
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: [1E6]]
Antibodies
SERPINA3 antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

PBS, pH 7.2

SERPINA3 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SERPINA3
Clonality: Monoclonal
Clone: [1E6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR