Anti-TFPI Mouse Monoclonal Antibody

Supplier: ProSci
51-663
10073-300EA 684.46 USD
10073-300
Anti-TFPI Mouse Monoclonal Antibody
Antibodies
TFPI antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation.

PBS, pH 7.2, 50% glycerol.

TFPI antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TFPI
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR