Anti-MUC16 Mouse Monoclonal Antibody [clone: [M002201]]

Supplier: ProSci
51-660
10073-294EA 540.97 USD
10073-294
Anti-MUC16 Mouse Monoclonal Antibody [clone: [M002201]]
CA-125 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

10mM Tris buffered saline with 0.05% NaN3

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUC16
Clonality: Monoclonal
Clone: [M002201]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR