Anti-CD34 Mouse Monoclonal Antibody [clone: [QBEnd/10]]

Supplier: ProSci
51-658
10073-292EA 540.97 USD
10073-292
Anti-CD34 Mouse Monoclonal Antibody [clone: [QBEnd/10]]
CD34 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

Azide-free PBS, pH 7.4; 0.2 micron filter sterilized

CD34 antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: [QBEnd/10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR