Anti-MARK3 Mouse Monoclonal Antibody

51-651
10073-278EA 540.97 USD InStock
10073-278
Anti-MARK3 Mouse Monoclonal Antibody
Antibodies
MARK3 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%

MARK3 antibody may be stored at 4 °C for short-term only. As with all antibodies avoid freeze/thaw cycles. Store at -20 °C for long term. Aliquots are stable for at least 12 months

Type:
Antigen: MARK3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR